Please login to your online account to display your discounted pricing

BD GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets


Used in producing anaerobic, microaerophilic, or CO2-enriched environments.   BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets provide catalyst-free convenience.

Manufacturer: bd  B260678

 View more versions of this product

 Disclaimers

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments

Catalog No. B260678

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Product Type GasPak™ EZ Anaerobe Container System Sachets
For Use With (Equipment) GasPak™ EZ anaerobe container system
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Storage Requirements 2° to 25°C
Format Sachet
Packaging 20 sachets/box

  • For use in both the GasPak™ EZ Containers and the GasPak™ 100 and GasPak™ 150 jars
  • Sachets require no water or catalyst for activation and are engineered to provide rapid gas generation for optimal bacterial growth