Please login to your online account to display your discounted pricing

BD GasPak™ EZ Gas Generating Systems Incubation Containers


Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.

Manufacturer: bd  B260672

 View more versions of this product

Catalog No. B260672

  • / Each
 

Description & Specifications

Specifications

Includes Removable Petridish racks
Storage Requirements 2 to 25°C
Closure Type Easy close latches
Disposable No
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Holds 30 to 33 petri dishes
Size Large
Product Type GasPak™ EZ Incubation Container

  • Non-breakable, chemical resistant containers