missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD GasPak™ 150 Systems

Vented and non-vented models

Manufacturer:  BD B260628

 View more versions of this product

Catalog No. 11-816



Includes: 10 1/2 O.D. × 11 in.H (27 × 28cm) polycarbonate jar; lid with O-ring gasket (vented or nonvented depending on model); clamp and thumbscrew assembly; three catalyst reaction chambers; 10 catalyst charges; autoclavable, stainless-steel Petri dish rack with clips; and culture tube holder. Vented model also supplied with rubber hose and tubing clamp.

Requires : Non-vented model (and vented model if tank system not used) requires disposable gas generator envelope.

Description

Description

Gaspak 150 Complete dimensions: 8.5 H x 10 in. dia. without the lid; 11.25 in. H with lid

Same as GasPak™ 100 Systems, but includes larger anaerobic jar

  • Provide all the components necessary to produce anaerobic, microaerophilic, or CO2-enriched aerobic atmospheres
  • Hold thirty-six 100 × 15mm Petri dishes, twelve 150 × 15mm Petri dishes, or 13 culture tubes
  • Both vented and non-vented models are available to meet a wide variety of needs

  • Non-vented model for anaerobic procedures
  • Vented model for evacuation-replacement systems that use tank gases

Specifications

Specifications

Generation of anaerobic, microaerophilic (reduced oxygen) or CO2-enriched environments
Polycarbonate jar (non-vented), lid with O-ring gasket, 3 catalyst reaction chambers, 10 charges, rack and tube holder
Large
Thirty six 100 x 15mm petri dishes, twelve 150 x 15mm petri dishes or 13 culture tubes
GasPak™ 150 Anaerobic System
No
SDS
Documents
Product Certifications
Promotions

Promotions