missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Filter Needle

Manufacturer:  BD 305200

Catalog No. 14-815-527



Description

Description

  • Helps reduce particulates larger than 5μ when aspirating, reconstituting and transferring fluids, or when withdrawing medication from glass ampules
  • BD Luer-Lok™ tip and molded-in membrane filter help eliminate the possibility of filter disassembly
  • Clear plastic hub permits continuous inspection of fluids
  • Large filter area provide high flow rate
  • Easy-to-peel package and fiber-free package materials

Order Info

Size: 19-gauge needle, 1 1/2 in. (3.8cm) long.

Specifications

Specifications

Thin Wall Filter Needle
19 G
Plastic
Clear
1.5 in.
Documents