missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD EnteroPluri Test

For detection of Enterobacteriaceae, other gram negative and oxidase negative bacteria

$447.72 - $447.72


Products 1
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
B215336 View Documents BD
B215336
Pack of 25 Each for $447.72
 
Description

Description

  • Disposable device used only for identification of bacteria from non-clinical samples
  • Allows simultaneous inoculation of all media present in sectors with execution of 15 biochemical reactions
  • Identification is based on biochemical tests performed on culture media with specific substrates
  • Change in color after 8 to 24 hours of incubation indicates presence of microorganisms
  • Consists of 12 sector system containing special culture media
    • Glucose/Gas: Glucose fermentation and gas production in anaerobiosis
    • Lysine: Lysine decarboxylation in anaerobiosis
    • Ornithine: Ornithine decarboxylation in anaerobiosis
    • H2S/ Indole: Hydrogen sulphide production and indole test
    • Adonitol: Adonitol fermentation
    • Lactose: Lactose fermentation
    • Arabinose: Arabinose fermentation
    • Sorbitol: Sorbitol fermentation
    • VP: Acetoin production (Voges-Proskauer)
    • Dulcitol/PA: Dulcitol fermentation and phenylalanine deamination
    • Urea: Urea hydrolysis
    • Citrate: Citrate utilization
  • For 25 tests

  • Codebook required for EnteroPluri Test helps in identification of bacteria