missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Eclipse Hypodermic Needles

Designed with pivoting needle technology for single-handed activation

Manufacturer:  BD B305763

 View more versions of this product

Catalog No. 14-821-22



Description

Description

No hard surface is required with this needle-based, safety-engineered injection device.

  • For use with BD Luer Lok
  • Sterile
  • Shelf Life: 5 years

Specifications

Specifications

Single-Handed
Luer-Lok
Minimize injection pain and patient discomfort with pivoting shielding mechanism, needle-based design facilitates standardization and SKU reduction
22g
Pivoting shielding mechanism
3.8cm
Regular
Eclipse™ Hypodermic Needle
Black
Yes
BD Luer-Lok syringe
Polypropylene
1.5 in.
Bevel point
Stainless Steel
Sterile
Documents