Please login to your online account to display your discounted pricing
BD Difco™ Salmonella O QC Antigens
Quick reactive Gold standard antisera-typing panels for Salmonella O antigen
Manufacturer: bd  B221371
 View more versions of this product
 Alert
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. DF2137-50-1
  • / Each
Description & Specifications
Specifications
For Use With (Application) Used in the quality control of Salmonella O Antisera Poly B (225351), Factor 7 (228161), Group C1 Factors 6 (240985), and Poly A-I & Vi (240993)
Product Type Group E4
Quantity 1mL
Shelf Life 730 days