Please login to your online account to display your discounted pricing

BD Difco™ Modified TSB Prepared Media with Casamino Acids

Used in qualitative procedures for the sterility test and for the enrichment and cultivation of aerobic microorganisms

$166.90 - $166.90

Specifications

Certifications/Compliance ISO/DIS, 11014, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Format Solid
For Use With (Application) Sterility testing and enrichment and cultivation of aerobic microorganisms
Packaging Tube
Manufacturing Origin United States
View More Specs
Products
Catalog Number Mfr. No. Media Type Quantity Price Quantity    

22-350-791

 
bd
B215237
Modified Trypticase Soy Broth with Casamino Acids 500g Each for $166.90
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Format Solid
For Use With (Application) Sterility testing and enrichment and cultivation of aerobic microorganisms
Packaging Tube
Manufacturing Origin United States
Shelf Life 1460 Days
Safety and Handling

1460 Days