missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Fluorescent Antibody Reagents
Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
$1,583.87 - $1,598.79
Specifications
For Use With (Application) | Direct fluorescent antibody technique for the identification of Bordetella pertussis and Bordetella parapertussis |
---|---|
Certifications/Compliance | ISO 13485:2003, ISO 9001:2008 |
Quantity | 5 mL |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
DF2359-56-6
|
BD
B223591 |
FA Bordetella Pertussis |
Each for $1,583.87
|
|
DF2378-56-3
|
BD
B223781 |
FA Bordetella Parapertussis |
Each for $1,598.79
|
|
Description
- Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
- For use in direct fluorescent identification
Order Info
In 5mL vials.
Specifications
Direct fluorescent antibody technique for the identification of Bordetella pertussis and Bordetella parapertussis | |
ISO 13485:2003, ISO 9001:2008 | |
5 mL |