BD Difco™ Fluorescent Antibody Reagents
Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
Manufacturer:  BD B223591
 View more versions of this product
Catalog No. DF2359-56-6
Description
Description
  • Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
  • For use in direct fluorescent identification
Order Info
In 5mL vials.
Specifications
Specifications
Black on Orange
Direct fluorescent antibody technique for the identification of Bordetella pertussis and Bordetella parapertussis
5 mL
ISO 13485:2003, ISO 9001:2008
FA Bordetella Pertussis