Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Yeast Nitrogen Base without Amino Acids and Ammonium Sulfate

Wickerham formula, classification of yeasts based on nitrogen and carbon requirements

$312.23 - $2,610.21

Specifications

Certifications/Compliance ISO/DIS, 11014
Format Dehydrated
For Use With (Application) For classifying Yeasts based on colonial characteristics and cell morphology
Media Type Yeast Nitrogen Base
Product Type Yeast Nitrogen base without Amino Acids and Ammonium Sulfate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DF0335-08-8

 
bd
B233510
Pail 10kg Each for $2,610.21

DF0335-15-9

 
bd
B233520
Bottle 100g Each for $312.23
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Format Dehydrated
For Use With (Application) For classifying Yeasts based on colonial characteristics and cell morphology
Media Type Yeast Nitrogen Base
Product Type Yeast Nitrogen base without Amino Acids and Ammonium Sulfate
Media Format Vitamins: Biotin 2μg/L, Calcium Pantothenate 400μg/L, Folic Acid 2μg/L, Inositol 2000μg/L, Niacin 400μg/L, p-Aminobenzoic Acid 200μg/L, Pyridoxine
Shelf Life 1825 Days
Storage Requirements <30°C

Prices are for units of one each unless otherwise specified.