Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Yeast Nitrogen Base without Amino Acids


Wickerham formula, classification of yeasts based on amino acid and carbohydrate requirements

Manufacturer: bd  B291930

 View more versions of this product

Catalog No. DF0919-08

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-8°C
Packaging Pail
Product Type Yeast Nitrogen without Amino Acids
Format Dehydrated
Quantity 10000g
Certifications/Compliance ISO, 9000
Media Type Yeast Nitrogen Base without Amino Acids
Shelf Life 1825 Days
For Use With (Application) For classifying Yeasts based on Nitrogen assimilation
Media Format Nitrogen Source: Ammonium Sulfate 5g/L, Vitamins: Biotin 2μg/L, Calcium Pantothenate 400μg/L, Folic Acid 2μg/L, Inositol 2000μg/L, Niacin 400μg/L, p-Aminobenzoic Aci
Accessories

1825 Days