Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: UVM Modified Listeria Enrichment Broth


Selective enrichment for culturing L. monocytogenes

Manufacturer: bd  B222320

 View more versions of this product

Catalog No. DF0223-08-3

  • / Each
 

Description & Specifications

Specifications

Packaging Pail
Certifications/Compliance AOAC, CCAM, COMPF, USDA
Quantity 10kg
Format Dehydrated
Media Format Pancreatic digest of Casein 5g/L, Proteose Peptone No. 3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 12mg/L
For Use With (Application) For rapidly isolating Listeria monocytogenes
Media Type Listeria Enrichment Broth
Shelf Life 1095 Days
Product Type UVM modified Listeria Enrichment broth
Storage Requirements Refrigerate
Accessories

1095 Days