Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: UVM Modified Listeria Enrichment Broth

Selective enrichment for culturing L. monocytogenes

$337.54 - $4,070.31

Specifications

Certifications/Compliance AOAC, CCAM, COMPF, USDA
Format Dehydrated
For Use With (Application) For rapidly isolating Listeria monocytogenes
Media Type Listeria Enrichment Broth
Product Type UVM modified Listeria Enrichment broth
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DF0223-07-4

 
bd
B222310
Pail 2kg Each for $861.85

DF0223-08-3

 
bd
B222320
Pail 10kg Each for $4,070.31

DF0223-17-2

 
bd
B222330
Bottle 500g Each for $337.54
Description & Specifications

Specifications

Certifications/Compliance AOAC, CCAM, COMPF, USDA
Format Dehydrated
For Use With (Application) For rapidly isolating Listeria monocytogenes
Media Type Listeria Enrichment Broth
Product Type UVM modified Listeria Enrichment broth
Media Format Pancreatic digest of Casein 5g/L, Proteose Peptone No. 3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 12mg/L
Shelf Life 1095 Days
Storage Requirements Refrigerate