Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Tryptose Blood Agar Base

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

$381.36 - $381.36

Specifications

Certifications/Compliance BAM
Format Dehydrated
For Use With (Application) For used with Blood in isolating cultivating and determining the hemolytic reactions
Media Type Tryptose Blood Agar Base
Product Type Tryptose Blood Agar base
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DF0232-17-1

 
bd
B223220
Bottle 500g Each for $381.36
Description & Specifications

Specifications

Certifications/Compliance BAM
Format Dehydrated
For Use With (Application) For used with Blood in isolating cultivating and determining the hemolytic reactions
Media Type Tryptose Blood Agar Base
Product Type Tryptose Blood Agar base
Media Format Tryptose 10g/L, Beef extract 3g/L, Sodium Chloride 5g/L, Agar 15g/L
Shelf Life 1825 Days

Used with blood for isolation and cultivation of fastidious microorganisms as well as determination of hemolytic reactions

BAM