missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Dehydrated Culture Media: Tryptose Blood Agar Base
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Supplier: BD 223220
Description
Used with blood for isolation and cultivation of fastidious microorganisms as well as determination of hemolytic reactions
Certifications
BAM
Specifications
BAM | |
Dehydrated | |
Agar | |
For used with Blood in isolating cultivating and determining the hemolytic reactions | |
500 g |
<30°C | |
Tryptose Blood Agar Base | |
Tryptose 10g/L, Beef extract 3g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Bottle |