Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Tryptic Soy Broth (Soybean-Casein Digest Medium)


Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Manufacturer: bd  B211822

 View more versions of this product

Catalog No. DF0370-07-5

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-25°C
Format Dehydrated
Media Format Pancreatic digest of Casein 17g/L, Papaic digest of Soybean 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L
Product Type Tryptic Soy broth (Soybean Casein digest medium)
Quantity 2000g
Shelf Life 1825 Days
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens
Media Type Tryptic Soy Broth
Packaging Pail
Certifications/Compliance AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP