Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar)


Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

Manufacturer: bd  B236940

 View more versions of this product

Catalog No. DF0369-15-8

  • / Each
 

Description & Specifications

Specifications

Packaging Bottle
Media Type Tryptic Soy Agar
Quantity 100g
Format Dehydrated
Product Type Tryptic Soy Agar(Soybean Casein digest Agar medium)
Certifications/Compliance AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Storage Requirements 2-30°C
For Use With (Application) Isolation of Escherichia coli 25922, Neisseria meningitidis 13090, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615, Aspergillus brasiliensis (niger) 16404, Bacillus subtillis 6633, Candida albicans 10231, Escherichia coli 8739, Pseudomonas aeruginosa 9027, Salmonella enterica subspecies enterica 14028, Staphylococcus aureus 6538 serotype Typhimurium
Shelf Life 1825 Days
Media Format Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L
Accessories

1825 Days