Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar with Lecithin and Polysorbate 80


Recommended for the detection and enumeration of microorganisms present on surfaces of sanitary importance

Manufacturer: bd  B255310

 View more versions of this product

Catalog No. DF0553-07-4

  • / Each
 

Description & Specifications

Specifications

Quantity 2kg
Certifications/Compliance CCAM
Product Type Microbial content test Agar
Format Dehydrated
Packaging Pail
Shelf Life 1460 Days
Media Type Microbial Content Test Agar
Media Format Pancreatic digest of Casein 15g/L, Soy Peptone 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
For Use With (Application) Detection and enumeration of microorganisms
Accessories

1460 Days