Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Tomato Juice Broth


For cultivation of yeasts and other aciduric microorganisms

Manufacturer: bd  B251720

 View more versions of this product

Catalog No. DF0517-17-7

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cultivation of Yeasts and other aciduric microorganisms
Format Dehydrated
Media Format Tomato Juice 20g/L, Yeast extract 10g/L, Dextrose 10g/L, Dipotassium Phosphate 0.5g/L, Monopotassium Phosphate 0.5g/L, Magnesium Sulfate 0.1g/L, Sodium Chloride 0.01g/L, Ferrous Sulfate 0.01g/L, Manganese Sulfate 0.01g/L
Media Type Tomato Juice Broth
Packaging Bottle
Product Type Tomato Juice broth
Quantity 500g
Shelf Life 1460 Days