Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Spirit Blue Agar

Used with Lipase Reagent for enumeration and detection of lipolytic microorganisms

$923.64 - $923.64

Specifications

Certifications/Compliance ISO/DIS, 11014, SMD
Format Dehydrated
For Use With (Application) Isolation, selective enrichment, differentiation, enumeration, estimation, maintenance, cultivation of bacteria
Media Type Spirit Blue Agar
Packaging Bottle
View More Specs
Products
Catalog Number Mfr. No. Storage Requirements Quantity Price Quantity    

DF0950-17-1

 
bd
B295020
25°C 500g Each for $923.64
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014, SMD
Format Dehydrated
For Use With (Application) Isolation, selective enrichment, differentiation, enumeration, estimation, maintenance, cultivation of bacteria
Media Type Spirit Blue Agar
Packaging Bottle
Product Type Spirit Blue Agar
Media Format Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Agar 20g/L, Spirit blue 0.15g/L
Shelf Life 1825 Days
Safety and Handling

1825 Days