Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Select APS™ Super Broth

To grow Escherichia coli to a high cell density

$206.72 - $2,051.18

Specifications

Certifications/Compliance CE
Format Dehydrated
For Use With (Application) Molecular Genetics medium used to grow Escherichia coli to a high cell density
Media Type Super Broth Base, Select APSI (Alternative Protein Source)
Product Type Super Broth
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B4312485

 
bd
B212485
Bottle 500g Each for $206.72

B4312486

 
bd
B212486
Pail 10kg Each for $2,051.18
Description & Specifications

Specifications

Certifications/Compliance CE
Format Dehydrated
For Use With (Application) Molecular Genetics medium used to grow Escherichia coli to a high cell density
Media Type Super Broth Base, Select APSI (Alternative Protein Source)
Product Type Super Broth
Media Format Soy Hydrolysate 12g/L, Yeast extract 24g/L, Dipotassium Phosphate 11.4g/L, Monopotassium Phosphate 1.7g/L
Shelf Life 1460 Days
Storage Requirements Refrigerate
Safety and Handling

1460 Days