Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Sabouraud Dextrose Broth


For cultivation of yeasts, molds and aciduric microorganisms

Manufacturer: bd  B238220

 View more versions of this product

Catalog No. DF0382-15-1

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-25°C
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Format Dehydrated
Media Format Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 20g/L
Media Type Sabouraud Dextrose Broth
Packaging Bottle
Product Type Sabouraud Dextrose broth
Quantity 100g
Shelf Life 1825 Days
Certifications/Compliance BAM, EP, JP, USP