Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Sabouraud Dextrose Broth

For cultivation of yeasts, molds and aciduric microorganisms

$87.28 - $312.69

Specifications

Certifications/Compliance BAM, EP, JP, USP
Format Dehydrated
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Media Type Sabouraud Dextrose Broth
Product Type Sabouraud Dextrose broth
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DF0382-07-1

 
bd
B238210
Pail 2000g Each for $312.69

DF0382-17-9

 
bd
B238230
Bottle 500g Each for $195.23

DF0382-15-1

 
bd
B238220
Bottle 100g Each for $87.28
Description & Specifications

Specifications

Certifications/Compliance BAM, EP, JP, USP
Format Dehydrated
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Media Type Sabouraud Dextrose Broth
Product Type Sabouraud Dextrose broth
Media Format Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 20g/L
Shelf Life 1825 Days
Shelf Life 1825 Days
Storage Requirements 2-25°C
Safety and Handling

1825 Days