Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Sabouraud Dextrose Agar


For cultivation of yeasts, molds and aciduric microorganisms

Manufacturer: bd  B210940

 View more versions of this product

Catalog No. DF0109-15-3

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-25°C
For Use With (Application) Cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes
Format Dehydrated
Media Type Sabouraud Dextrose Agar
Packaging Bottle
Product Type Sabouraud Dextrose Agar
Quantity 100g
Certifications/Compliance BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Media Format Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 40g/L, Agar 15g/L
Shelf Life 1825 Days