Please login to your online account to display your discounted pricing

BD™ BD™ Difco™ R2A Agar


Intended for use with stressed and chlorine-tolerant bacteria. BD™ Difco™ R2A Agar is a dehydrated, low-nutrient medium for enumerating and cultivating compromised bacteria and other heterotrophic organisms. It was developed for bacteriological plate counts of treated potable water.

Manufacturer: bd™  B218262

 View more versions of this product

Catalog No. DF1826-15-3

  • / Each
 

Description & Specifications

Specifications

Format Dehydrated (free flowing, homogeneous)
Color Light beige
Quantity 100g
Product Type R2A Agar
Certifications/Compliance COMPF, EPA, SMWW
Packaging Bottle
For Use With (Application) Low nutrient medium for enumeration and cultivation of bacteria from potable Water
Media Format Yeast extract 0.5g/L, Proteose Peptone No. 3 0.5g/L, Casamino Acids 0.5g/L, Dextrose 0.5g/L, Soluble Starch 0.5g/L, Sodium Pyruvate 0.3g/L, Dipotassium Phosphate 0.3g/L, Magnesium Sulfate 0.05g/L, Agar 15g/L
Media Type R2A Agar
Shelf Life 1825 Days

R2A Agar was developed for bacteriological plate counts of treated potable water. A low nutrient medium, such as R2A Agar, in combination with a lower incubation temperature and longer incubation time stimulates the growth of stressed and chlorine-tolerant bacteria.

Nutritionally-rich media, such as Plate Count Agar, support the growth of fast-growing bacteria but may suppress slow growing or stressed bacteria found in treated water. When compared with nutritionally rich media, R2A Agar has been reported to improve the recovery of stressed and chlorine-tolerant bacteria from drinking water systems.

  • Recommended in standard methods for pour plate, spread plate and membrane filter methods for heterotrophic plate counts.
  • Intended for use only with compromised bacteria found in treated potable water.
  • Yeast extract provides a source of trace elements and vitamins.
  • Peptone and casamino acids provide nitrogen, vitamins, amino acids, carbon and minerals.
  • Dextrose serves as a carbon source.
  • Soluble starch aids in the recovery of injured organisms by absorbing toxic metabolic by-products.
  • Sodium pyruvate increases the recovery of stressed cells.
  • Potassium phosphate is used to balance the pH and provide phosphate.
  • Magnesium sulfate is a source of divalent cations and sulfate.
  • Agar is the solidifying agent.

Certifications

COMPF, EPA, SMWW

Accessories

1825 Days