Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: mFC Broth Base


For cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperature

Manufacturer: bd  B288320

 View more versions of this product

Catalog No. DF0883-15-5

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance CE, EPA, SMWW
Format Dehydrated
Packaging Bottle
Storage Requirements Refrigerate
Media Type mFC Broth Base
Quantity 100g
Shelf Life 1460 Days
Product Type m FC Broth Base
For Use With (Application) Used with Rosolic Acid in cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperature
Media Format Tryptose 10g/L, Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Aniline blue 0.1g/L
Accessories

1460 Days