Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Membrane Fecal Coliform (mFC) Agar


For detection and enumeration of fecal coliforms by the membrane filter technique at elevated temperatures

Manufacturer: bd  B267710

 View more versions of this product

Catalog No. DF0677-15-5

  • / Each
 

Description & Specifications

Specifications

Packaging Bottle
Quantity 100g
Storage Requirements Refrigerate
Media Type mFC Agar
Product Type Membrane Fecal Coliform Agar
Certifications/Compliance AOAC, CCAM, EPA, SMWW
Format Dehydrated
For Use With (Application) For Rosolic Acid in cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperatures
Media Format Tryptose 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts No. 3 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Aniline blue 0.1g/L
Shelf Life 1460 Days
Accessories

1460 Days