Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: m Enterococcus Agar


For isolating and enumerating enterococci in water and other materials by membrane filtration or pour plate technique

Manufacturer: bd  B274610

 View more versions of this product

Catalog No. DF0746-15-2

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For isolating and enumerating enterococci in Water and other materials by membrane filtration or pour plate technique
Format Dehydrated
Media Format Tryptose 20g/L, Yeast extract 5g/L, Dextrose 2g/L, Dipotassium Phosphate 4g/L, Sodium Azide 0.4g/L, Agar 10g/L, 2,3,5-Triphenyl Tetrazolium Chloride 0.1g/L
Media Type m Enterococcus Agar
Packaging Bottle
Product Type m Enterococcus Agar
Quantity 100g
Shelf Life 1825 Days
Storage Requirements <30°C
Certifications/Compliance CE, ISO, SMWW