Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: m Endo Broth MF™


For enumerating coliform organisms in water by membrane filtration

Manufacturer: bd  B274920

 View more versions of this product

Catalog No. DF0749-15-9

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Millipore™ filter, one step membrane filtration technique for the enumeration of Coliforms
Format Dehydrated
Media Format Yeast extract 1.5g Casitone 5g/L, Thiopeptone 5g/L, Tryptose 10g/L, Lactose 12.5g/L, Sodium Desoxycholate 0.1g/L, Dipotassium Phosphate 4.375g/L, Monopotassium Phosphate 1.375g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 2.1g/L, Basic Fuchsin 1.05g/L
Packaging Bottle
Product Type m Endo broth MF™
Storage Requirements <30°C
Certifications/Compliance COMPF, SMD, SMWW
Media Type m Endo Broth MF-Millipore
Quantity 100g
Shelf Life 1825 Days