Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: m E Agar

Used with nalidixic acid and triphenyltetrazolium chloride in isolating and differentiating enterococci from water by membrane filtration and in an in situ esculin test on Esculin Iron Agar

$392.51 - $392.51

Specifications

Certifications/Compliance ISO, 9000, EPA, SMWW
Format Dehydrated
For Use With (Application) Isolation of Enterococcus faecalis 29212, Enterococcus faecalis 33186, Escherichia coli 25922
Media Type mE Agar
Packaging Bottle
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

DF0333-17-9

 
bd
B233320
500g Each for $392.51
Description & Specifications

Specifications

Certifications/Compliance ISO, 9000, EPA, SMWW
Format Dehydrated
For Use With (Application) Isolation of Enterococcus faecalis 29212, Enterococcus faecalis 33186, Escherichia coli 25922
Media Type mE Agar
Packaging Bottle
Product Type m E Agar
Media Format Yeast extract 30g/L, Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Agar 15g/L
Shelf Life 1460 Days
Storage Requirements <30°C

  • Formulation: Yeast extract 30g/L, Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Agar 15g/L

ISO, 9000, EPA, SMWW