Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Luria Agar Base, Miller


For propagation and maintenance of E. coli for use in molecular biology

Manufacturer: bd  B211829

 View more versions of this product

Catalog No. DF0413-07-4

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance CE
For Use With (Application) Maintaining and propagating Escherichia coli in molecular microbiology procedures with or without added Glucose
Format Dehydrated
Media Format Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Sodium Chloride 0.5g/L, Agar 15g/L
Media Type Luria Agar Base
Packaging Pail
Product Type Luria Agar Base
Quantity 2kg
Shelf Life 1825 Days
Storage Requirements Refrigerate
Accessories

1825 Days