Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Luria Agar Base, Miller

For propagation and maintenance of E. coli for use in molecular biology

$350.10 - $740.46

Specifications

Certifications/Compliance CE
Format Dehydrated
For Use With (Application) Maintaining and propagating Escherichia coli in molecular microbiology procedures with or without added Glucose
Media Type Luria Agar Base
Product Type Luria Agar Base
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DF0413-07-4

 
bd
B211829
Pail 2kg Each for $740.46

DF0413-17-2

 
bd
B241320
Bottle 500g Each for $350.10
Description & Specifications

Specifications

Certifications/Compliance CE
Format Dehydrated
For Use With (Application) Maintaining and propagating Escherichia coli in molecular microbiology procedures with or without added Glucose
Media Type Luria Agar Base
Product Type Luria Agar Base
Media Format Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Sodium Chloride 0.5g/L, Agar 15g/L
Shelf Life 1825 Days
Storage Requirements Refrigerate

  • Contains 0.5g/L NaCl

Applications:

For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria