Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Listeria Enrichment Broth

Selective enrichment for L. monocytogenes from non-dairy and processed food products

$251.38 - $2,642.36

Specifications

Format Dehydrated
For Use With (Application) Selectively enrich Listeria from food
Media Type Listeria Enrichment Broth
Product Type Listeria Enrichment broth
Media Format Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Yeast extract 6g/L, Cycloheximide 0.05g/L, Acriflavine HCl 15mg/L, Nalidixic Acid 0.04g/L
View More Specs
Products
Catalog Number Mfr. No. Certifications/Compliance Packaging Quantity Price Quantity    

DF0222-08-4

 
bd
B222210
ISO 9001:2008 Pail 10kg Each for $2,642.36

DF0222-17-3

 
bd
B222220
CE Bottle 500g Each for $251.38
Description & Specifications

Specifications

Format Dehydrated
For Use With (Application) Selectively enrich Listeria from food
Media Type Listeria Enrichment Broth
Product Type Listeria Enrichment broth
Media Format Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Yeast extract 6g/L, Cycloheximide 0.05g/L, Acriflavine HCl 15mg/L, Nalidixic Acid 0.04g/L
Shelf Life 1460 Days
Storage Requirements Refrigerate

For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

1460 Days