BD Difco™ Dehydrated Culture Media: Listeria Enrichment Broth
Selective enrichment for L. monocytogenes from non-dairy and processed food products
Manufacturer:  BD B222210
 View more versions of this product
Catalog No. DF0222-08-4
Specifications Description & Specifications
Specifications
ISO 9001:2008
Dehydrated
Listeria Enrichment Broth
Listeria Enrichment broth
Refrigerate
Selectively enrich Listeria from food
Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Yeast extract 6g/L, Cycloheximide 0.05g/L, Acriflavine HCl 15mg/L, Nalidixic Acid 0.04g/L
Pail
10kg