Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: LB Agar, Miller (Luria-Bertani)


Propagate and maintain E. coli for molecular biology.  BD Difco™ Dehydrated Culture Media: LB Agar, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

Manufacturer: bd  B244520

 View more versions of this product

Catalog No. DF0445-17-4

  • / Each
 

Description & Specifications

Specifications

Product Type LB Agar
Media Type LB Agar, Miller
Certifications/Compliance CCAM
For Use With (Application) Propagation and maintenance of Escherichia coli for molecular biology
Format Dehydrated
Media Format Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 15g/L
Packaging Bottle
Quantity 500g
Shelf Life 1825 Days
Storage Requirements <30°C

 

  • Contains 10g/L NaCl
  • For propagation of E. coli
  • Wide range of molecular biology applications