Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Lauryl Tryptose Broth


Medium for detection of coliform bacteria

Manufacturer: bd  B224140

 View more versions of this product

Catalog No. DF0241-15-2

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance CE, AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW
For Use With (Application) Detection of Coliform organisms in materials of sanitary importance
Format Dehydrated
Media Format Tryptose 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L
Media Type Lauryl Tryptose Broth
Packaging Bottle
Product Type Lauryl Tryptose broth
Quantity 100g
Shelf Life 1825 Days
Storage Requirements Refrigerate
Accessories

1825 Days