Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Lauryl Tryptose Broth with MUG


Broth with MUG Fluorogenic detection of E. coli in water, food and milk

Manufacturer: bd  B211740

 View more versions of this product

Catalog No. DF0021-15-8

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance CE, AOAC, BAM, COMPF, SMD, SMWW
Quantity 100g
Format Dehydrated
Packaging Bottle
For Use With (Application) Detection of Escherichia coli in Water, food and dairy samples by a fluorogenic procedure
Media Format Tryptose 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L, 4-methylumbelliferyl-beta-D-glucuronide 0.05g/L
Storage Requirements <30°C
Product Type Lauryl Tryptose broth with 4-methylumbelliferyl-beta-D-glucuronide
Media Type Lauryl Tryptose Broth
Shelf Life 1095 Days
Accessories

1095 Days