Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Lauryl Tryptose Broth with MUG

Broth with MUG Fluorogenic detection of E. coli in water, food and milk

$112.33 - $859.26

Specifications

Certifications/Compliance CE, AOAC, BAM, COMPF, SMD, SMWW
Format Dehydrated
For Use With (Application) Detection of Escherichia coli in Water, food and dairy samples by a fluorogenic procedure
Media Format Tryptose 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L, 4-methylumbelliferyl-beta-D-glucuronide 0.05g/L
Shelf Life 1095 Days
View More Specs
Products
Catalog Number Mfr. No. Media Type Packaging Quantity Price Quantity    

DF0021-15-8

 
bd
B211740
Lauryl Tryptose Broth Bottle 100g Each for $112.33

DF0021-17-6

 
bd
B211744
Lauryl Tryptose Broth Bottle 500g Each for $859.26
Description & Specifications

Specifications

Certifications/Compliance CE, AOAC, BAM, COMPF, SMD, SMWW
Format Dehydrated
For Use With (Application) Detection of Escherichia coli in Water, food and dairy samples by a fluorogenic procedure
Media Format Tryptose 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L, 4-methylumbelliferyl-beta-D-glucuronide 0.05g/L
Shelf Life 1095 Days
Storage Requirements <30°C
Safety and Handling

1095 Days