Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: L-Arabinose


Manufacturer: bd  B215910

 View more versions of this product

Catalog No. DF0159-13-4

  • / Each
 

Description & Specifications

Specifications

Packaging Bottle
Quantity 25g
Certifications/Compliance ISO/DIS, 11014
Shelf Life 1825 Days
Format Solid
Storage Requirements <30°C
Additive L-Arabinose
For Use With (Application) Microbiological culture media in the study of fermentation reactions
Accessories

1825 Days