Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: L-Arabinose

$238.82 - $612.15

Specifications

Additive L-Arabinose
Certifications/Compliance ISO/DIS, 11014
Format Solid
For Use With (Application) Microbiological culture media in the study of fermentation reactions
Shelf Life 1825 Days
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

DF0159-13-4

 
bd
B215910
25g Each for $238.82

DF0159-15-2

 
bd
B215920
100g Each for $612.15
Description & Specifications

Specifications

Additive L-Arabinose
Certifications/Compliance ISO/DIS, 11014
Format Solid
For Use With (Application) Microbiological culture media in the study of fermentation reactions
Shelf Life 1825 Days
Safety and Handling

1825 Days