Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Granulated Agar


Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.

Manufacturer: bd  B214520

 View more versions of this product

Catalog No. DF0145-08-0

  • / Each
 

Description & Specifications

Specifications

Concentration 1.5%
Additive Agar
For Use With (Application) General bacteriological purposes
Format Dehydrated Granule
Packaging Container
Quantity 10000g
Shelf Life 1825 Days
Certifications/Compliance ISO/DIS, 11014