Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium


For sterility testing of biologics and cultivation of anaerobes, aerobes and microaerophiles

Manufacturer: bd  B225640

 View more versions of this product

Catalog No. DF0256-15-4

  • / Each
 

Description & Specifications

Specifications

Format Dehydrated
Product Type Fluid Thioglycollate medium
Packaging Bottle
Quantity 100g
Shelf Life 1825 Days
Certifications/Compliance AOAC, BAM, BS12, COMPF, EP, USDA, USP
Media Type Fluid Thioglycollate Medium
For Use With (Application) Used for sterility testing
Media Format Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Storage Requirements 2-8°C
Accessories

1825 Days