Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium with Beef Extract

Used in cultivating microorganisms from normally sterile biological products

$272.41 - $272.41

Specifications

Certifications/Compliance CE, USP
Format Dehydrated
For Use With (Application) Cultivation of microorganisms from normally sterile biological product
Product Type Fluid Thioglycollate medium with Beef extract
Media Format Beef extract 5g/L, Yeast extract 5g/L, Pancreatic digest of Casein 15g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DF0697-17-9

 
bd
B269720
Bottle 500g Each for $272.41
Description & Specifications

Specifications

Certifications/Compliance CE, USP
Format Dehydrated
For Use With (Application) Cultivation of microorganisms from normally sterile biological product
Product Type Fluid Thioglycollate medium with Beef extract
Media Format Beef extract 5g/L, Yeast extract 5g/L, Pancreatic digest of Casein 15g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Shelf Life 1825 Days
Storage Requirements Refrigerate

  • Recommended by the Agricultural Research Service
  • Formulation: Beef extract 5g/L, Yeast extract 5g/L, Pancreatic digest of Casein 15g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L

CE, USP