Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Fluid Tetrathionate


Used as a selective enrichment medium for the isolation of Salmonella

Manufacturer: bd  B210430

 View more versions of this product

Catalog No. DF0104-17-6

  • / Each
 

Description & Specifications

Specifications

Packaging Bottle
Quantity 500g
Media Type Fluid Tetrathionate Medium
Shelf Life 730 Days
Product Type Tetrathionate broth base
Format Dehydrated
Storage Requirements <30°C
Certifications/Compliance AOAC, BAM, CCAM, COMPF, SMD, SMWW
For Use With (Application) Isolation of Salmonella from feces, urine, foods and other materials of sanitary importance
Media Format Proteose Peptone 2.5g/L, Pancreatic digest of Casein 2.5g/L, Oxgall 1g/L, Sodium Thiosulfate 30g/L, Calcium Carbonate 10g/L
Accessories

730 Days