Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Fluid Tetrathionate


Used as a selective enrichment medium for the isolation of Salmonella

Manufacturer: bd  B210420

 View more versions of this product

Catalog No. DF0104-07-8

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO, 9000, AOAC, BAM, CCAM, COMPF, SMD, SMWW
For Use With (Application) Isolation of Escherichia coli 25922, Salmonella enterica subspecies enterica serotype Typhimurium 14028
Format Dehydrated
Media Format Proteose Peptone 2.5g/L, Pancreatic digest of Casein 2.5g/L, Oxgall 1g/L, Sodium Thiosulfate 30g/L, Calcium Carbonate 10g/L
Media Type Fluid Tetrathionate Medium
Packaging Pail
Product Type Fluid Tetrathionate Medium
Quantity 2000g
Shelf Life 730 Days
Storage Requirements <30°C
Accessories

730 Days