Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Fluid Lactose Medium


For cultivation of Salmonella and coliform organisms in water, foods, dairy and pharmaceutical products

Manufacturer: bd  B243000

 View more versions of this product

Catalog No. DF0004-15-9

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-30°C
Packaging Bottle
Format Dehydrated
Shelf Life 1825 Days
Certifications/Compliance ISO, 9000, AOAC, BAM, CCAM, COMPF, SMD
Product Type Fluid Lactose medium
Quantity 100g
For Use With (Application) Cultivation of Salmonella and Coliform organisms in Water, foods, dairy and pharmaceutical products
Media Type Lactose Broth
Media Format Beef extract 3g/L, Peptone 5g/L, Lactose 5g/L
Accessories

1825 Days