Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: EC Medium, Modified

Used with Novobiocin Antimicrobic Supplement in the detection of Escherichia coli O157:H7 in meat and poultry products

$205.44 - $205.44

Specifications

Certifications/Compliance CCAM, COMPF
Format Dehydrated
Media Type EC Medium
Product Type EC Medium, Modified
Media Format Pancreatic digest of Casein 20g/L, Bile Salts No. 3 1.12g/L, Lactose 5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Packaging Quantity Price Quantity    

DF0340-17-0

 
bd
B234020
For Novobiocin antimicrobic supplement in the detection of Escherichia coli O157:H7 in meat and poultry products Bottle 500g Each for $205.44
Description & Specifications

Specifications

Certifications/Compliance CCAM, COMPF
Format Dehydrated
Media Type EC Medium
Product Type EC Medium, Modified
Media Format Pancreatic digest of Casein 20g/L, Bile Salts No. 3 1.12g/L, Lactose 5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L
Shelf Life 1825 Days
Storage Requirements <30°C

  • Formulation: Pancreatic digest of Casein 20g/L, Bile Salts No. 3 1.12g/L, Lactose 5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L
    • CCAM, COMPF

Safety and Handling

1825 Days