Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: EC medium with 4-methylumbelliferyl-beta-D-glucuronide


For detecting Escherichia coli in water, food and milk

Manufacturer: bd  B222200

 View more versions of this product

Catalog No. DF0022-17-5

  • / Each
 

Description & Specifications

Specifications

Shelf Life 1095 Days
Quantity 500g
Storage Requirements <30°C
For Use With (Application) Detection Escherichia coli in Water, food and Milk
Media Type E.Coli Medium
Packaging Bottle
Certifications/Compliance BAM, CCAM, EPA, SMWW
Format Dehydrated
Product Type EC medium with 4-methylumbelliferyl-beta-D-glucuronide
Media Format Tryptose 20g/L, Lactose 5g/L, Bile Salts No. 3 1.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L, 4-methylumbelliferyl-beta-D-glucuronide 0.05g/L
Accessories

1095 Days