Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: EC medium with 4-methylumbelliferyl-beta-D-glucuronide

For detecting Escherichia coli in water, food and milk

$437.38 - $922.00

Specifications

Certifications/Compliance BAM, CCAM, EPA, SMWW
Format Dehydrated
For Use With (Application) Detection Escherichia coli in Water, food and Milk
Media Type E.Coli Medium
Packaging Bottle
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

DF0022-15-7

 
bd
B222100
100g Each for $437.38

DF0022-17-5

 
bd
B222200
500g Each for $922.00
Description & Specifications

Specifications

Certifications/Compliance BAM, CCAM, EPA, SMWW
Format Dehydrated
For Use With (Application) Detection Escherichia coli in Water, food and Milk
Media Type E.Coli Medium
Packaging Bottle
Product Type EC medium with 4-methylumbelliferyl-beta-D-glucuronide
Media Format Tryptose 20g/L, Lactose 5g/L, Bile Salts No. 3 1.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L, 4-methylumbelliferyl-beta-D-glucuronide 0.05g/L
Shelf Life 1095 Days
Storage Requirements <30°C

  • Formulation: Tryptose 20g/L, Lactose 5g/L, Bile Salts No. 3 1.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L, 4-methylumbelliferyl-beta-D-glucuronide 0.05g/L

BAM, CCAM, EPA, SMWW

Safety and Handling

1095 Days